Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-12
Entrez Gene ID
192074
Gene Name
TWiK family of potassium channels protein 12
Gene Aliases
CELE_F29F11.4
Chromosome Location
Chr.V: 10667584 - 10670890 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073330.5 NP_505731.3 6 - 7 760 91

View Full Product Details