Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-22
Entrez Gene ID
185095
Gene Name
Serpentine Receptor, class J
Gene Aliases
CELE_F28H7.1
Chromosome Location
Chr.V: 10742423 - 10743749 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073338.2 NP_505739.1 5 - 6 829 80

View Full Product Details