Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
nrx-1
Entrez Gene ID
179503
Gene Name
protein-coding
Gene Aliases
CELE_C29A12.4
Chromosome Location
Chr.V: 10825011 - 10846703 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129396.5 NP_001122868.2 9 - 10 1121 129
NM_001269333.4 NP_001256262.1 9 - 10 1124 129

View Full Product Details