Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sec-61.G
Entrez Gene ID
179510
Gene Name
Protein transport protein Sec61 subunit gamma
Gene Aliases
CELE_F32D8.6
Chromosome Location
Chr.V: 10896656 - 10897149 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383391.1 1 - 2 101 70

View Full Product Details