Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-74
Entrez Gene ID
183468
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C45B11.4
Chromosome Location
Chr.V: 11047220 - 11048778 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073412.2 NP_505813.1 1 - 2 249 121

View Full Product Details