Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lact-8
Entrez Gene ID
189839
Gene Name
protein-coding
Gene Aliases
CELE_Y42A5A.2
Chromosome Location
Chr.V: 11091016 - 11093619 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269357.2 NP_001256286.1 5 - 6 946 121
NM_001269358.3 NP_001256287.1 5 - 6 672 121
NM_001269359.3 NP_001256288.1 3 - 4 324 121

View Full Product Details