Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-49
Entrez Gene ID
179547
Gene Name
Ligand-Gated ion Channel
Gene Aliases
CELE_K10D6.1
Chromosome Location
Chr.V: 11191283 - 11193639 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073440.4 NP_505841.2 3 - 4 388 135

View Full Product Details