Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-29A2
Entrez Gene ID
179550
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_T19B10.1
Chromosome Location
Chr.V: 11217583 - 11219911 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073446.5 NP_505847.1 2 - 3 333 70

View Full Product Details