Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-18
Entrez Gene ID
179565
Gene Name
CUTiclin-Like
Gene Aliases
CELE_F47G9.3
Chromosome Location
Chr.V: 11313362 - 11316901 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073474.6 NP_505875.2 2 - 3 565 80

View Full Product Details