Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grl-7
Entrez Gene ID
179569
Gene Name
protein-coding
Gene Aliases
CELE_T02E9.2
Chromosome Location
Chr.V: 11344825 - 11347012 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028922.4 NP_001024093.1 2 - 3 543 60

View Full Product Details