Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccch-1
Entrez Gene ID
179584
Gene Name
protein-coding
Gene Aliases
CELE_F38B7.1
Chromosome Location
Chr.V: 11527831 - 11541219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073525.8 NP_505926.2 9 - 10 1342 118
NM_073526.5 NP_505927.2 7 - 8 1387 118

View Full Product Details