Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-90
Entrez Gene ID
186725
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H12C20.5
Chromosome Location
Chr.V: 11581126 - 11582458 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073537.1 NP_505938.1 5 - 6 760 145

View Full Product Details