Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pyc-1
Entrez Gene ID
179616
Gene Name
protein-coding
Gene Aliases
CELE_D2023.2
Chromosome Location
Chr.V: 11817783 - 11822580 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383412.1 6 - 7 1810 75
GenBank mRNA AF237467.1 - 5 - 6 1733 75

View Full Product Details