Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hmp-1
Entrez Gene ID
179624
Gene Name
protein-coding
Gene Aliases
CELE_R13H4.4
Chromosome Location
Chr.V: 11845871 - 11852176 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129519.3 NP_001122991.1 2 - 3 839 110
NM_001276937.3 NP_001263866.1 2 - 3 838 110
NM_001373121.3 NP_001360176.1 2 - 3 845 110
NM_001383414.1 2 - 3 839 110
NM_073584.6 NP_505985.1 2 - 3 839 110
GenBank mRNA AF016852.1 - 2 - 3 861 110

View Full Product Details