Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hmp-1
Entrez Gene ID
179624
Gene Name
protein-coding
Gene Aliases
CELE_R13H4.4
Chromosome Location
Chr.V: 11845871 - 11852176 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129519.3 NP_001122991.1 6 - 7 2550 103
NM_073584.6 NP_505985.1 6 - 7 2550 103
GenBank mRNA AF016852.1 - 6 - 7 2572 103

View Full Product Details