Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sulp-5
Entrez Gene ID
187335
Gene Name
STAS domain-containing protein
Gene Aliases
CELE_K12G11.2
Chromosome Location
Chr.V: 11880032 - 11883219 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073589.5 NP_505990.3 6 - 7 959 141
GenBank mRNA AY887911.1 - 6 - 7 981 141

View Full Product Details