Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sodh-1
Entrez Gene ID
179627
Gene Name
Alcohol dehydrogenase 1
Gene Aliases
CELE_K12G11.3
Chromosome Location
Chr.V: 11888236 - 11892673 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073590.7 NP_505991.1 2 - 3 882 100

View Full Product Details