Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aman-2
Entrez Gene ID
179632
Gene Name
Alpha-mann_mid domain-containing protein
Gene Aliases
CELE_F58H1.1
Chromosome Location
Chr.V: 11927661 - 11933246 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073594.4 NP_505995.2 6 - 7 1571 73

View Full Product Details