Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gcy-33
Entrez Gene ID
179644
Gene Name
protein-coding
Gene Aliases
CELE_F57F5.2
Chromosome Location
Chr.V: 11995805 - 12001797 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269462.4 NP_001256391.1 8 - 9 2507 82
NM_001269463.3 NP_001256392.1 8 - 9 2492 82
NM_001269464.3 NP_001256393.1 1 - 2 374 82
GenBank mRNA AY275181.1 - 8 - 9 2486 82

View Full Product Details