Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gana-1
Entrez Gene ID
179660
Gene Name
Alpha-galactosidase
Gene Aliases
CELE_R07B7.11
Chromosome Location
Chr.V: 12087066 - 12088935 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073630.8 NP_506031.1 5 - 6 1142 95

View Full Product Details