Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vab-8
Entrez Gene ID
179675
Gene Name
protein-coding
Gene Aliases
CELE_K12F2.2
Chromosome Location
Chr.V: 12213327 - 12225938 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129505.5 NP_001122977.1 6 - 7 1710 70
NM_073662.6 NP_506063.2 14 - 15 3066 70
NM_073663.6 NP_506064.1 5 - 6 1838 70

View Full Product Details