Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ugt-55
- Entrez Gene ID
-
188072
- Gene Name
- Putative UDP-glucuronosyltransferase ugt-55
- Gene Aliases
-
CELE_T04H1.7
- Chromosome Location
-
Chr.V: 12256274 - 12258804 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_073672.6 |
NP_506073.2 |
8 - 9 |
1470 |
99 |
- Gene Symbol
- T04H1.12
- Entrez Gene ID
-
36805035
- Gene Name
- Uncharacterized protein
- Gene Aliases
-
CELE_T04H1.12
- Chromosome Location
-
Chr.V: 12255547 - 12258793 on Build WBcel235
View Full Product Details