Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tbb-6
Entrez Gene ID
179680
Gene Name
Tubulin beta chain
Gene Aliases
CELE_T04H1.9
Chromosome Location
Chr.V: 12261531 - 12263368 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073674.4 NP_506075.2 2 - 3 251 73

View Full Product Details