Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-16
Entrez Gene ID
186287
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_F55C5.9
Chromosome Location
Chr.V: 12291861 - 12294196 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073683.3 NP_506084.2 7 - 8 1134 129

View Full Product Details