Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-45
Entrez Gene ID
183710
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_C51F7.2
Chromosome Location
Chr.V: 12307490 - 12308658 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073685.1 NP_506086.1 2 - 3 364 116

View Full Product Details