Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-13
Entrez Gene ID
192077
Gene Name
protein-coding
Gene Aliases
CELE_R04F11.4
Chromosome Location
Chr.V: 12320170 - 12337994 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129512.4 NP_001122984.1 6 - 7 1102 83
NM_001276939.4 NP_001263868.1 8 - 9 857 83
NM_001276940.3 NP_001263869.1 7 - 8 700 83
NM_073690.10 NP_506091.2 6 - 7 631 83

View Full Product Details