Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gcy-13
Entrez Gene ID
191646
Gene Name
Receptor-type guanylate cyclase gcy-13
Gene Aliases
CELE_F23H12.6
Chromosome Location
Chr.V: 12363754 - 12369537 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073696.3 NP_506097.3 14 - 15 2660 87

View Full Product Details