Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-14
Entrez Gene ID
179699
Gene Name
protein-coding
Gene Aliases
CELE_K01D12.4
Chromosome Location
Chr.V: 12387039 - 12395706 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001276941.4 NP_001263870.1 6 - 7 943 108
NM_001380784.1 NP_001367281.1 6 - 7 908 108
NM_001380785.1 NP_001366791.1 6 - 7 921 108

View Full Product Details