Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdr-4
Entrez Gene ID
259654
Gene Name
CaDmium Responsive
Gene Aliases
CELE_K01D12.11
Chromosome Location
Chr.V: 12407366 - 12409146 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073709.6 NP_506110.1 3 - 4 312 107
GenBank mRNA AY728064.1 - 2 - 3 283 107

View Full Product Details