Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdr-2
Entrez Gene ID
183779
Gene Name
CaDmium Responsive
Gene Aliases
CELE_C54D10.1
Chromosome Location
Chr.V: 12414622 - 12415709 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073713.4 NP_506114.1 3 - 4 435 120
GenBank mRNA AY728062.1 - 3 - 4 457 120

View Full Product Details