Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdr-3
Entrez Gene ID
183780
Gene Name
CaDmium Responsive
Gene Aliases
CELE_C54D10.2
Chromosome Location
Chr.V: 12416358 - 12417377 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073714.2 NP_506115.2 4 - 5 687 152

View Full Product Details