Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-25
Entrez Gene ID
183783
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C54D10.6
Chromosome Location
Chr.V: 12432365 - 12433817 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073718.4 NP_506119.4 4 - 5 831 96

View Full Product Details