Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-10
Entrez Gene ID
179707
Gene Name
protein-coding
Gene Aliases
CELE_F28C1.2
Chromosome Location
Chr.V: 12453290 - 12463008 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269489.4 NP_001256418.1 2 - 3 421 85
NM_001269490.3 NP_001256419.1 2 - 3 229 85
NM_001269491.2 NP_001256420.1 2 - 3 421 85
GenBank mRNA U32326.1 - 2 - 3 427 85

View Full Product Details