Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-39
Entrez Gene ID
189186
Gene Name
protein-coding
Gene Aliases
CELE_W04D2.2
Chromosome Location
Chr.V: 12484885 - 12486068 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047710.1 NP_001041175.1 4 - 5 704 74
NM_001322589.1 NP_001309669.1 2 - 3 581 74
NR_003176.1 2 - 3 447 74

View Full Product Details