Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
inx-11
Entrez Gene ID
179710
Gene Name
protein-coding
Gene Aliases
CELE_W04D2.3
Chromosome Location
Chr.V: 12487234 - 12493269 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269497.3 NP_001256426.1 1 - 2 201 84

View Full Product Details