Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-43
Entrez Gene ID
179750
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_F18E2.4
Chromosome Location
Chr.V: 12769246 - 12771018 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073793.5 NP_506194.3 1 - 2 268 137

View Full Product Details