Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-43
Entrez Gene ID
191176
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_ZC455.8
Chromosome Location
Chr.V: 12789197 - 12790518 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047739.1 NP_001041204.1 2 - 3 536 169

View Full Product Details