Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-5
Entrez Gene ID
179753
Gene Name
protein-coding
Gene Aliases
CELE_ZC455.6
Chromosome Location
Chr.V: 12790677 - 12792965 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073799.5 NP_506200.1 7 - 8 1226 72
NM_182346.5 NP_872146.1 7 - 8 1116 72

View Full Product Details