Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-6
Entrez Gene ID
179755
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_ZC455.4
Chromosome Location
Chr.V: 12795484 - 12798154 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383430.1 5 - 6 1195 69

View Full Product Details