Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-16
Entrez Gene ID
191171
Gene Name
UDP-glucuronosyltransferase
Gene Aliases
CELE_ZC443.6
Chromosome Location
Chr.V: 12825024 - 12827442 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073809.7 NP_506210.1 3 - 4 482 111

View Full Product Details