Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-28
Entrez Gene ID
187889
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_R90.3
Chromosome Location
Chr.V: 12904213 - 12905174 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073825.4 NP_506226.2 2 - 3 315 136

View Full Product Details