Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-68
Entrez Gene ID
188555
Gene Name
D-aminoacyl-tRNA deacylase
Gene Aliases
CELE_T16G1.10
Chromosome Location
Chr.V: 12952636 - 12953208 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047703.2 NP_001041168.1 2 - 3 261 68
NR_003175.1 2 - 3 255 68

View Full Product Details