Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
far-4
Entrez Gene ID
191633
Gene Name
Fatty Acid/Retinol binding protein
Gene Aliases
CELE_F15B9.2
Chromosome Location
Chr.V: 12998427 - 12999659 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073851.5 NP_506252.1 1 - 2 430 89

View Full Product Details