Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
him-17
Entrez Gene ID
179800
Gene Name
High Incidence of Males (increased X chromosome loss)
Gene Aliases
CELE_T09E8.2
Chromosome Location
Chr.V: 13176711 - 13180189 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073876.4 NP_506277.1 5 - 6 1076 105

View Full Product Details