Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-15
Entrez Gene ID
183634
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_C50B6.6
Chromosome Location
Chr.V: 13320976 - 13322381 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073901.3 NP_506302.2 5 - 6 867 118

View Full Product Details