Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-4
Entrez Gene ID
183637
Gene Name
Seven TM Receptor
Gene Aliases
CELE_C50B6.10
Chromosome Location
Chr.V: 13337904 - 13339733 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073906.2 NP_506307.1 7 - 8 955 123

View Full Product Details