Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpz-2
Entrez Gene ID
179818
Gene Name
Pept_C1 domain-containing protein
Gene Aliases
CELE_M04G12.2
Chromosome Location
Chr.V: 13381189 - 13383077 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073917.7 NP_506318.1 4 - 5 843 99

View Full Product Details