Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-8
Entrez Gene ID
186171
Gene Name
Serpentine Receptor, class V
Gene Aliases
CELE_F53F1.7
Chromosome Location
Chr.V: 13418951 - 13420740 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073926.3 NP_506327.2 4 - 5 513 97

View Full Product Details