Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-19
Entrez Gene ID
191808
Gene Name
Serpentine receptor class delta-19
Gene Aliases
CELE_F53F1.11
Chromosome Location
Chr.V: 13427607 - 13428760 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073930.1 NP_506331.1 1 - 2 192 97

View Full Product Details