Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-17
Entrez Gene ID
191806
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_Y2H9A.2
Chromosome Location
Chr.V: 13430651 - 13432714 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073931.1 NP_506332.1 2 - 3 426 117

View Full Product Details